Oxaliplatin

Supplier: MORAVEK BIOCHEMICALS MS

MC2321-10UCI MC2321-50UCI
101971-398EA 1693.46 USD
101971-398 101971-400
Oxaliplatin
Oxaliplatin
Formula: C₈H₁₄N₂O₄Pt
MDL Number: MFCD00866327
CAS Number: 61825-94-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR