Oleyl alcohol, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2303-50UCI MC2303-250UCI
101971-366EA 5387.69 USD
101971-366 101971-368
Oleyl alcohol, [1-14C] ≥98%, MORpure™
Oleyl alcohol, [1-14C]
CAS Number: 2088576-32-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR