N6-Benzyladenine, [benzyl ring-14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2302-50UCI MC2302-250UCI
101971-362EA 5387.69 USD
101971-362 101971-364
N6-Benzyladenine, [benzyl ring-14C(U)] ≥98%, MORpure™
N6-Benzyladenine, [benzyl ring-14C(U)]
CAS Number: 2088617-21-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR