8,11,14-Eicosatrienoic acid, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2212-10UCI MC2212-50UCI
101971-186EA 1837.31 USD
101971-186 101971-188
8,11,14-Eicosatrienoic acid, [1-14C] ≥98%, MORpure™
8,11,14-Eicosatrienoic acid, [1-14C]
CAS Number: 3435-80-1
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR