Acetic anhydride, [1-14C] ≥95% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC2188-1MCI MC2188-5MCI
101971-128EA 8192.69 USD
101971-128 101971-126
Acetic anhydride, [1-14C] ≥95% (by HPLC)
Acetic anhydride, [1-14C]
CAS Number: 344322-57-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR