Adenosine, [8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC151-10UCI MC151-50UCI MC151-250UCI
101970-682EA 1693.46 USD
101970-682 101970-684 101970-686
Adenosine, [8-14C] ≥98%, MORpure™
Adenosine, [8-14C]
CAS Number: 3257-92-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR