Nicotinamide, [carbonyl-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1427-250UCI MC1427-50UCI
101970-548EA 6826.15 USD
101970-548 101970-546
Nicotinamide, [carbonyl-14C] ≥98% (by HPLC), MORpure™
Nicotinamide, [carbonyl-14C]
CAS Number: 10119-18-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR