Isopropanol, [2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1161-250UCI MC1161-50UCI
101970-168EA 5191.54 USD
101970-168 101970-170
Isopropanol, [2-14C] ≥97% (by HPLC)
Isopropanol, [2-14C]
CAS Number: 104810-30-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR