L-Valine, [1-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1133-50UCI MC1133-250UCI
101970-132EA 5387.69 USD
101970-132 101970-134
L-Valine, [1-14C] ≥98% (by HPLC), MORpure™
L-Valine, [1-14C]
CAS Number: 14330-62-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR