Thymidine, [2-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC104-10UCI MC104-50UCI MC104-250UCI
101970-074EA 1693.46 USD
101970-074 101970-076 101970-078
Thymidine, [2-14C] ≥98%, MORpure™
Thymidine, [2-14C]
CAS Number: 13010-45-2
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR