Suramin hexasodium

Supplier: MORAVEK BIOCHEMICALS MS

M823-1MG M823-5MG
101970-020EA 294.23 USD
101970-020 101970-022
Suramin hexasodium
Suramin hexasodium
Formula: C₅₁H₃₄N₆Na₆O₂₃S₆
MDL Number: MFCD00210217
CAS Number: 129-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR