Paclitaxel

Supplier: MORAVEK BIOCHEMICALS MS

M552-10MG M552-25MG
101969-944EA 294.23 USD
101969-944 101969-946
Paclitaxel
Paclitaxel
Formula: C₄₇H₅₁NO₁₄
MDL Number: MFCD00869953
CAS Number: 33069-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR