1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)uracil ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-((2R,3S,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil

M439-1MG M439-5MG
101969-924EA 294.23 USD
101969-924 101969-926
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)uracil ≥98%, MORpure™
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)uracil
Formula: C₉H₁₁FN₂O₅
MW: 246.1925 g/mol
MDL Number: MFCD00870798
CAS Number: 69123-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR