β-L-Cytidine ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-1-((2S,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, L-Cytidine

M320-1MG M320-5MG
101969-888EA 294.23 USD
101969-888 101969-890
β-L-Cytidine ≥98%, MORpure™
β-L-Cytidine
Formula: C₉H₁₃N₃O₅
MW: 243.2166 g/mol
MDL Number: MFCD02683605
CAS Number: 26524-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR