Foscarnet sodium ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: Phosphonoformate, Sodium phosphonatoformate, Trisodium phosphonoformate, Phosphonoformic acid trisodium salt

M305-1MG M305-5MG
101969-884EA 294.23 USD
101969-884 101969-886
Foscarnet sodium ≥98%, MORpure™
Foscarnet sodium
Formula: CNa₃O₅P
MW: 191.95 g/mol
MDL Number: MFCD00013973
CAS Number: 63585-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR