2'-Methylcytidine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

M1864-1MG M1864-5MG
101969-752EA 294.23 USD
101969-752 101969-754
2'-Methylcytidine ≥98% (by HPLC) MORpure™
2'-Methylcytidine
Formula: C₁₀H₁₅N₃O₅
MDL Number: MFCD02682947
CAS Number: 20724-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR