7-Deazaadenosine ≥98%, MORpure™
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
4-Amino-7-(β-D-ribofuranosyl)pyrrolo[2,3-d]pyrimidine
M1808-1MG
M1808-5MG
101969-672EA
294.23
USD
101969-672
101969-674
7-Deazaadenosine ≥98%, MORpure™
7-Deazaadenosine
Formula:
C₁₁H₁₄N₄O₄ MW: 266.2533 g/mol |
MDL Number:
MFCD00056012 CAS Number: 69-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...