8-Bromoguanosine
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one
M1793-1MG
M1793-5MG
101969-608EA
294.23
USD
101969-608
101969-610
8-Bromoguanosine
8-Bromoguanosine
Formula:
C₁₀H₁₂BrN₅O₅ MW: 362.14 g/mol |
MDL Number:
MFCD00037985 CAS Number: 4016-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...