2'-Fluoro-2'-deoxycytidine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-1-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, 2'-Deoxy-2'-fluorocytidine

M1787-1MG M1787-5MG
101969-588EA 294.23 USD
101969-588 101969-590
2'-Fluoro-2'-deoxycytidine
2'-Fluoro-2'-deoxycytidine
Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
MDL Number: MFCD00057445
CAS Number: 10212-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR