5-Methylcytidine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one

M1784-1MG M1784-5MG
101969-576EA 294.23 USD
101969-576 101969-578
5-Methylcytidine
5-Methylcytidine
Formula: C₁₀H₁₅N₃O₅
MW: 257.25 g/mol
MDL Number: MFCD00006548
CAS Number: 2140-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR