5-Aza-2'-deoxycytidine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

M1774-1MG M1774-5MG
101969-540EA 294.23 USD
101969-540 101969-542
5-Aza-2'-deoxycytidine
5-Aza-2'-deoxycytidine
Formula: C₈H₁₂N₄O₄
MDL Number: MFCD00043011
CAS Number: 2353-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR