3'-(O-Methyl)-adenosine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)-4-methoxytetrahydrofuran-3-ol3’-O-Methyladenosine

M1741-1MG M1741-5MG
101969-426EA 738.85 USD
101969-426 101969-424
3'-(O-Methyl)-adenosine ≥98% (by HPLC) MORpure™
3'-(O-Methyl)adenosine
Formula: C₁₁H₁₅N₅O₄
MW: 281.2679 g/mol
MDL Number: MFCD00057003
CAS Number: 10300-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR