Emtricitabine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-5-fluoro-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

M1605-1MG M1605-5MG
101969-306EA 294.23 USD
101969-306 101969-308
Emtricitabine
Emtricitabine
Formula: C₈H₁₀FN₃O₃S
MW: 247.24 g/mol
MDL Number: MFCD00870151
CAS Number: 143491-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR