tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate ≥98%
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
tert-Butyl bis(2-hydroxyethyl)carbamate, N-BOC-Diethanolamine
12904-5ML
12904-25ML
12904-100ML
102567-062EA
110.35
USD
102567-062
102567-064
102621-226
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate ≥98%
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate
Synonyms: Tert-Butyl N,N-bis(2-hydroxyethyl)carbamate
Formula:
C₉H₁₉NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD01862953 CAS Number: 103898-11-9 |
Specification Test Results
Appearance | Light yellow clear liquid |
Solubility | Soluble in Methanol |
Purity Limit | ≥ 98% (Assay) |
1H NMR | Conforms to structure |
Refractive Index | 1.46 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...