LaMotte® Lead Screening Test
Supplier: LaMotte
470326-588EA 136.99 USD
470326-588
LaMotte® Lead Screening Test
Test Paper Indicator Paper
In only 10 minutes the analyst can detect lead levels above 15 ppb- the EPA action level.
  • Includes 10 tests that come individually sealed with test vial and dropper
  • Results in 10 minutes
  • Detects lead to EPA action level of 15 ppb
Simply immerse the strip in a water sample and read after a 10 minute waiting period.
The multi-test package includes 10 tests and diagrammed instuctions.