Tetrabutylammonium chloride ≥98%

Supplier: CHEM IMPEX INTL INC MS

27306-1KG 27306-100G 27306-25G 27306-250G
102584-942EA 36.83 USD
102584-942 102560-842 102560-840 102604-526
Tetrabutylammonium chloride ≥98%
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
MW: 277.92 g/mol
MDL Number: MFCD00011635
CAS Number: 1112-67-0

Order Now

Specification Test Results

Appearance White to off-white crystalline powder
Assay ≥ 99% (Anhydrous basis)
Storage Temperature 0 - 8 °C
Water Content ≤ 3%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR