RediSep® Disposable Sample Load Cartridges, Teledyne ISCO
Supplier: Teledyne
693873235 693873240 693873224
101663-922PK 178.46 USD
101663-922 101663-930 101671-824
RediSep® Disposable Sample Load Cartridges, Teledyne ISCO
Chromatography Columns
These empty disposable sample load cartridges improve the resolution of the compound and eliminate reaction byproducts when compared to liquid injection techniques.
Prepare pre-filled solid load cartridges by pipetting the dissolved sample onto the top of the cartridge. Prepare empty solid load cartridges by filling the cartridge with a slurry mixture of the dissolved sample and supporting media. For optimal benefits, remove the solvent by vacuum before placing the cartridge on the purification system.