BD Taxo™ ALA Discs, BD Diagnostic
Supplier: BD Diagnostics
BD Taxo™
231725 231725
90002-546PK 105.21 USD
CA90002-546 90002-546
BD Taxo™ ALA Discs, BD Diagnostic
Microbiology Test Systems Microorganism Tests
BD Taxo™ Differentiation Discs ALA are used for rapidly detecting porphyrin as a means of speciating Haemophilus species.
These are paper discs that contain buffered delta-aminolevulinic acid.