Agar, Calbiochem®
Supplier: MilliporeSigma
Synonyms:
Agar Agar
12177-1KG
EM12177-1KGEA
344.26
USD
EM12177-1KG
Agar, Calbiochem®
Agar
Suitable for inclusion in culture media.
Storage and Shipping Instructions: Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 month at 4°C.
Storage Temperature:
Ambient |
MDL Number:
MFCD00081288 CAS Number: 9002-18-0 Merck Index: 14, 184 |
Specification Test Results
Form | Beige solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...