Potassium chloride USP, JP, Ph. Eur., BP for bioprocessing, suitable for use as excipient endotoxin tested

Supplier: Avantor Performance Materials

Synonyms: Potassiumchloride

VWRB395-12KG VWRB395-500G
VWRB395-12KGEA 929.63 USD
VWRB395-12KG VWRB395-2.5KG VWRB395-500G
Potassium chloride USP, JP, Ph. Eur., BP for bioprocessing, suitable for use as excipient endotoxin tested
Potassium chloride

  • Tested according to the USP, EP, BP and JP monographs
  • Material is of non-animal, non-human origin.
  • BSE/TSE free.
  • Endotoxin tested.


Packaging: Custom packaging and testing available. Change management and notification available. Samples available.

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Merck Index: 14,07621

Order Now

Specification Test Results

Acidity or Alkalinity (USP, EP, BP, JP) PASS
Assay (dried basis, USP) 99.0 - 100.5%
Assay (dried substance, EP, BP) 99.0 – 101.0%
Assay (dried, JP) ≥ 99.0%
Bacterial Endotoxins ≤ 2.5 EU/g
Arsenic <1.11> ≤ 2 ppm
Barium Test (EP, BP) PASS
Bromides (USP, EP, BP) ≤ 0.1%
Bromide Test (JP) PASS
Calcium and Magnesium Test (USP, JP) PASS
Clarity and color of solution PASS
Heavy Metals <231> ≤ 10 ppm
Heavy Metals <1.07> ≤ 5 ppm
Identification A (USP, EP, BP) PASS
Identification B (USP, EP, BP) PASS
Identification (JP) PASS
Iodide (USP) ≤ 0.005%
Iodides Test (EP, BP, JP) PASS
Iron (2.4.9) ≤ 20 ppm
Loss on Drying <731>, 2.2.32 ≤ 1.0%
Loss on Drying <2.41> ≤ 1.0%
Sodium Test (USP, JP) PASS
Sulfates (2.4.13) ≤ 300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR