S-Monovette® Blood Collecting System, Sarstedt
Supplier: Sarstedt
S-Monovette®
05.1167.100 06.1664.100 04.1901.100 04.1931.100
101433-390PK 23.37 USD
101433-390 101093-936 101093-948 101094-006
S-Monovette® Blood Collecting System, Sarstedt
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
S-Monovette® is a blood collection system that combines the aspiration technique and the vacuum technique.
  • Aspiration technique as a gentle technique for routine blood collection
  • Using the vacuum technique, a "fresh" vacuum is always available
S-Monovette® Hematology EDTA K3 is pre-dosed as a liquid preparation in an average concentration of 1.6 mg EDTA/ml blood. The maximum dilution caused by the liquid preparation is lower than 1%. An S-Monovette® with EDTA K2 and gel is available for use in molecular virus diagnostics.
Certifications: CE certified