Mitis Salivarius Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M259-500G M259-500G
95021-570EA 299.52 USD
95021-570 CA95021-570
Mitis Salivarius Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation of streptococci from mixed cultures, especially Streptococcus mitis, Streptococcus salivarius, and Streptococcus faecalis.
  • Microbiological culture media
  • 500 g poly bottles