Diaphragm Vacuum Pump, D-LAB, Edwards
Supplier: Edwards
A48022941 A48016941 A48018941 A48021941 A48015941
EVA480-22-941EA 7401.36 USD
EVA480-22-941 EVA480-16-941 EVA480-18-941 EVA480-21-941 EVA480-15-941
Diaphragm Vacuum Pump, D-LAB, Edwards
Pumps Vacuum Pumps
D-LAB Diaphragm Vacuum Pumps are lightweight and compact design which is ideal for chemical or pharmaceutical laboratory use with a wide range of choice.
  • Safe, easy to transport and plug in connectors
  • Chemically resistant Teflon® heads
  • High ultimate vacuum and fast pump down time
  • Gas-tight, 100 oil-free, maintenance-free and quiet
  • Operate independently or as part of a modular system
  • Valve systems are tolerant of water vapour and condensates
Illuminated switch on pump and overload protection by means of thermal cut-out
The one head, 100 mbarMore Product Information