Orange Serum Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M933-500G M933-500G
95022-972EA 173.19 USD
CA95022-972 95022-972
Orange Serum Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and enumeration of microorganisms associated with spoilage of citrus products, and for the cultivation of lactobacilli and other aciduric organisms and pathogenic fungi.
  • Microbiological culture media
  • 500 g poly bottles