Anti-Kappa Immunoglobulin Light Chain Mouse Monoclonal Antibody [clone: MAR 18.5]

Supplier: Bio X Cell
BE0122-1MG BE0122-100MG BE0122-5MG BE0122-50MG BE0122-25MG
103253-788EA 3920 USD
103253-788 103253-790 103253-792 103253-794 103572-812 76554-686 76555-092 76555-802 76556-858 76556-862
Anti-Kappa Immunoglobulin Light Chain Mouse Monoclonal Antibody [clone: MAR 18.5]
Antibodies
InVivoMab anti-rat Kappa Immunoglobulin Light Chain, Clone: MAR 18.5, Isotype: mouse IgG2a, κ, Applications: in vivo administration, Flow cytometry

Type: Primary
Antigen: Kappa Immunoglobulin Light Chain
Clonality: Monoclonal
Clone: MAR 18.5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR