Anti-TIM-4 Rat Monoclonal Antibody [clone: RMT4-54]

Supplier: Bio X Cell
BE0225-1MG BE0225-50MG BE0225-25MG BE0225-5MG BE0225-100MG
103253-088EA 3920 USD
103253-088 103253-090 103253-092 103253-094 103572-576 76555-918 76556-600 76556-698 76556-744 76557-152
Anti-TIM-4 Rat Monoclonal Antibody [clone: RMT4-54]
Antibodies
InVivoMab anti-mouse TIM-4, Clone: RMT4-54, Isotype: rat IgG2a, Applications: in vitro TIM-4 blocking, Immunofluorescence, Flow cytometry

Type: Primary
Antigen: TIM-4
Clonality: Monoclonal
Clone: RMT4-54
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR