Anti-CD48 Armenian Hamster Monoclonal Antibody [clone: HM48-1]

Supplier: Bio X Cell
BE0147-50MG BE0147-1MG BE0147-25MG BE0147-100MG BE0147-5MG
103252-812EA 3920 USD
103252-812 103252-814 103252-816 103252-818 103572-548 76554-358 76555-548 76555-828 76556-212 76557-308
Anti-CD48 Armenian Hamster Monoclonal Antibody [clone: HM48-1]
Antibodies
InVivoMab anti-mouse CD48, Clone: HM48-1, Isotype: Armenian hamster IgG, Applications: in vivo CD48 blockade, in vitro CD48 blocking

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: HM48-1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR