RediSep Rf Gold® C18Aq Chromatography Column, Teledyne ISCO
Supplier: Teledyne
692203560 692203562
101671-890EA 379.08 USD
101671-890 101671-894
RediSep Rf Gold® C18Aq Chromatography Column, Teledyne ISCO
Chromatography Columns
RediSep Rf Gold C18Aq is designed for highly aqueous conditions.
  • Monofunction C18 bonding is interspersed with hydrophilic ligands
  • Prevents phase collapse in high aqueous conditions
  • Used in separations requiring 0 to 50% organic
These columns are useful for highly polar, water soluble compounds such as dyes, glycopeptides, and nucleotides.