Glucose Salt Teepol Broth, Twin Pack, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M621-500G M621S-500G
95022-094EA 301.48 USD
95022-094 95022-096
Glucose Salt Teepol Broth, Twin Pack, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the enrichment of Vibrio parahaemolyticus and marine isolates from foods
  • Microbiological culture media
  • 500 g poly bottles