Haemophilus Test Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1259-500G M1259-500G
95020-808EA 325.46 USD
CA95020-808 95020-808
Haemophilus Test Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For susceptibility testing of Haemophilus influenzae
  • Microbiological culture media
  • 500 g poly bottles