Sodium acetate ≥99.0% (by titrimetric analysis) for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Acetic acid sodium salt
71185-250G
71185-50G
BJ71185-250GEA
549
USD
BJ71185-250G
BJ71185-50G
Sodium acetate ≥99.0% (by titrimetric analysis) for HPLC, Fluka™
Sodium acetate
Formula:
H₃CCOONa Boiling Pt: Decomposes on heating Melting Pt: 324 °C Density: 1.45 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 |
Specification Test Results
Assay (HClO₄-titr.) | 99.0 - 101.0 % |
Absorbance at 250 nm | Max. 0.05 |
Absorbance at 260 nm | Max. 0.01 |
Absorbance 500 nm | Max. 0.01 |
Appearance of the solution (c=10%) | complying |
pKa | 4.76 (acetic acid) |
Form | Solid |
Autoignition Temperature | 1112 °F |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...