Ketogluconate Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1324-500G M1324-500G
95020-936EA 210.44 USD
CA95020-936 95020-936
Ketogluconate Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For identifying bacteria that can utilize [alpha]-ketogluconate.
  • Microbiological culture media
  • 500 g poly bottles