Karmali Campylobacter Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1222-500G M1222-2.5KG M1222-500G M1222-5KG M1222-5KG M1222-2.5KG
95020-730EA 216.4 USD
CA89223-398 CA95020-730 95020-730 89223-396 CA89223-396 89223-398
Karmali Campylobacter Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and cultivation of thermotolerant Campylobacter species from food and animal feeds.
  • Microbiological culture media