Litmus Milk Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M635-500G M609-2.5KG M609-500G M609S-5KG M609S-5KG M609-5KG M609S-2.5KG M609S-2.5KG M609S-500G M609-500G M609-2.5KG M609S-500G M609-5KG
89225-988EA 604.41 USD
89225-988 CA89225-986 CA89225-988 CA61000-124 61000-124 89225-990 CA95022-110 89225-984 89225-986 CA89225-984 CA95022-062 CA89225-990 95022-062
Litmus Milk Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the maintenance of Lactobacilli and for determining the action of bacteria on milk.
  • Microbiological culture media
  • pH: 6.8 ±0.2
Certifications: M609S - recommended by BIS committee under specifications IS:5887(Part-IV)-1976.