Bolton Broth Base, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1592-500G M1592-2.5KG M1592-5KG
95021-316EA 266.91 USD
95021-316 89224-338 89224-340
Bolton Broth Base, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Campylobacter species from food and animal foodstuffs.
  • Microbiological culture media
  • pH: 7.20 - 7.60