Ayers and Johnson Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M182-500G M182-500G
95021-460EA 283.01 USD
95021-460 CA95021-460
Ayers and Johnson Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the maintenance of cultures of streptococci and other microorganisms
  • Microbiological culture media
  • Packaged in 500 g poly bottles