EC Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M127I-500G M127-500G GM127-2.5KG GM127-500G GM127-5KG
95020-856EA 128.01 USD
95020-856 95020-836 75804-082 75804-084 75804-086
EC Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
EC Broth is recommended for the selective enumeration of presumptive E. coli by MPN technique from water samples and from clinical samples.
  • Microbiological culture media
Certifications: Complies as per ISO 17025:2005.