SHOWA S-TEX303 A6 ANSI Cut Resistant, Wrinkle Finish, Palm Dipped, Kevlar/Steel Glove, Showa
Supplier: Showa
S-TEX303S-07 S-TEX303L-09 S-TEX303XL-10
10852-310CS 2498.58 USD
10852-310 10852-312 10852-314 10852-316
SHOWA S-TEX303 A6 ANSI Cut Resistant, Wrinkle Finish, Palm Dipped, Kevlar/Steel Glove, Showa
Gloves
Extemely high level protection while maintaining excellent flexibility.
  • A6 ANSI cut level
  • Kevlar/Hagane steel construction
  • Ergonomic design
Use where added protection from cuts is required. Wrinkle finish and ergonimic design provide added grip and dexterity.