SHOWA KV660 Chemical Resistant/Cut Resistant, 12 Gauntlet Glove, Showa
Supplier: Showa
KV660XXL-11 KV660XL-10
89208-326CS 1910.58 USD
89208-326 89208-320 89208-324
SHOWA KV660 Chemical Resistant/Cut Resistant, 12 Gauntlet Glove, Showa
Gloves
A flexible easy-maintenance glove providing effective protection against cuts and with very good durability.
  • Surface provides tactile feel and better grip
  • Scalloped edge
  • Seamless knit
  • Cut level 3, PVC-dipped, resistant to oil and hydrocarbons
Seals and protects the hand against chemicals while remaining flexible, impermeable for working in damp or greasy environments, enabling you to grip objects securely.