Jar Rolling Mills, Bench, Sepor
Supplier: SEPOR INC
010E-027 010E-035 110V/60HZ
80700-004EA 7926.85 USD
80700-004 80700-006
Jar Rolling Mills, Bench, Sepor
Mills
These continuous duty jar drives are used for batch wet or dry grinding, blending, dispersing or other conditioning.
  • Roll spacing is adjustable to accommodate jar sizes of 2 to 15" dia.
  • All models have free wheeling jar stops
  • Constructed of highest quality materials
Variable speed DC drive for an output range of 20 to 345 roller RPM; thus an 8 inch diameter jar would have a speed range of 5 to 86 RPM. Water tight electrical system and totally enclosed gear motor. These mill drives are built to last with very little maintenance and can drive heavy,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable